Learn More
Abeba™ Arrow 8300 Shoes
Antistatic with heel patch microfiber. Sanitized™ treated.
Brand:  Abeba™ 8300/45
Additional Details : Weight : 0.60000kg
 View more versions of this product
Product Code. 15835081
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
  • CE EN ISO 20347:2012 OB FO SRA
  • Microfiber
  • White
  • Lightweight Mono-PU-outsole
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
  • Microfiber heel patch
Specifications
Specifications
CE EN ISO 20347:2012 OB FO SRA
Microfiber
8300
White
45