Learn More
Abeba™ Arrow 8300 Shoes
Antistatic with heel patch microfiber. Sanitized™ treated.
59.45€
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 OB FO SRA |
---|---|
Color | White |
Material | Microfiber |
Style | 8300 |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15845071
|
Abeba™
8300/36 |
36 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
15855071
|
Abeba™
8300/37 |
37 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
15865071
|
Abeba™
8300/38 |
38 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
15875071
|
Abeba™
8300/39 |
39 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
15885071
|
Abeba™
8300/40 |
40 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
15895071
|
Abeba™
8300/41 |
41 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
15805081
|
Abeba™
8300/42 |
42 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
15815081
|
Abeba™
8300/43 |
43 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
15825081
|
Abeba™
8300/44 |
44 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
15835081
|
Abeba™
8300/45 |
45 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
15845081
|
Abeba™
8300/46 |
46 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
15855081
|
Abeba™
8300/47 |
47 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Description
- CE EN ISO 20347:2012 OB FO SRA
- Microfiber
- White
- Lightweight Mono-PU-outsole
- Sanitized treated
- Antistatic
- Heel strap foldable, adjustable
- Microfiber heel patch
Specifications
CE EN ISO 20347:2012 OB FO SRA | |
White | |
Microfiber | |
8300 |