Learn More
Abeba™ Arrow 38200 Shoes
Product Code. 15611138
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15611138 36 1 pair
15621138 37 1 pair
15631138 38 1 pair
15641138 39 1 pair
15651138 40 1 pair
15661138 41 1 pair
15671138 42 1 pair
15681138 43 1 pair
15691138 44 1 pair
15601148 45 1 pair
15611148 46 1 pair
15621148 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 15611138 Supplier Abeba™ Supplier No. 38200/36
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Antistatic with heel patch microfiber. Sanitized™ treated.
  • CE EN ISO 20347:2012 OB A, FO SRA
  • Smooth leather white
  • Slip resistance PU/PU outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • Microfiber heel patch
  • Lining microfiber, respiration absorbing
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRA
Color White
Liner Material Microfiber
Material Leather
Size 36
Sole Features Slip-resistant
Sole Material Polyurethane
Style 38200