missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ARMET Recombinant Protein

Brand:  Abnova™ H00007873-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192172

  • 335.00€ / 10µg
Estimated Shipment: 28-08-2024
to see stock.
Add to basket

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH07282.1
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
46
8
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ARMET
Human
Recombinant
Solution
Antibody Production, Protein Array, ELISA, Western Blot
7873
ARMET (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
ARP/MANF/MGC142148/MGC142150
ARMET
Wheat Germ (in vitro)
GST
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers