missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Arium™ Tubing Extension Set

Product Code. p-7221162 Shop All Sartorius Products
Click to view available options
For Use With (Equipment):
Arium Smart Station Ultrapure Water
Arium™ Smart Station Pure Water
Unit Size:
1 unit
This item is not returnable. View return policy

Product Code. 17204233

Brand: Sartorius H2OATESP

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Extension set (4 m) for Arium™ Smart Station, used for uncomplicated extension of connection between Arium™ Smart Station and connected water system

  • Extended tubing set used for Smart Station ultrapure and pure for uncomplicated extension of standard supplied tubing set
  • Operating range can be easily increased from 2 to 4 m
TRUSTED_SUSTAINABILITY

Specifications

Color (Overall) White
For Use With (Equipment) Arium™ Smart Station Pure Water
Type Tubing
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.