Learn More
Sartorius Arium™ Smart Station UP
Shop All Sartorius Products
Click to view available options
Type:
Remote Dispenser
Remote Dispenser Support
Unit Size:
1 unit
Description
- Height adjustable display/dispense station for dispensing Type 1 ultrapure water
- Compact design for easy and space-saving integration into lab environment
- Intuitive touch-activated color display with direct access to important functions
- Flexible and stepless height adjustment to fill different sized containers and beakers
- Ergonomic design can be optimized for right or left hand operation
- Accurate volume dispense for fast and precise buffer and sample prep
- Adjustable, splash-free flow rate from drop-by-drop to 2 L/min
Equipment supplied:
- Display/dispense unit with stand
- Power supply including:
- IEC connector
- Connection set
For intended use with:
- Arium™ Bagtank
- Arium™ Pro
- Arium™ Comfort
Specifications
Specifications
| Capacity (Metric) | 50 L |
| Feed Water Monitoring | No |
| Feedwater Source | Ultrapure |
| Flow Rate | Up to 2 L/min. |
| Product Water Type | Ultrapure (Type 1) |
| TOC Monitor with UV Intensity Monitoring | No |
| Type | Remote Dispenser Support |
| Display | Touch display |