missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ARHGEF7 Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.25 mg
Unit Size:
250µg
Specifications
Specifications
| Gene ID (Entrez) | 8874 |
| Species | Human |
| Concentration | 2.5 mg/mL |
| Content And Storage | Store at -20°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP3-12453. |
| Gene Alias | Beta-Pix, COOL1COOL-1, DKFZp686C12170, DKFZp761K1021, guanine nucleotide exchange factor 7, KIAA0142KIAA0412, Nbla10314, P50, P50BP, P85, P85COOL1, P85SPRBETA-PIX, PAK3, PAK3BP, PIXBPAK-interacting exchange factor beta, Rho guanine nucleotide exchange factor (GEF) 7, rho guanine nucleotide exchange factor 7, SH3 domain-containing proline-rich protein |
| Gene Symbol | ARHGEF7 |
| Quantity | 0.25 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction