Learn More
Tocris Bioscience™ Argatroban
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent inhibitor of thrombin mediated fibrinogen cleavage (Ki = 19 nM). Competitive inhibitor of thrombin-induced platelet activation and clotting. Shown to exhibit antithrombotic activity in animal models.
This compound is a mixture of 2 diastereomers (approximate ratio 65/35).
Specifications
Specifications
| Chemical Name or Material | Argatroban |
| CAS | 74863-84-6 |
| Assay Percent Range | >99% |
| Quantity | 10 mg |
| Molecular Formula | C23H36N6O5S |
| Formula Weight | 508.63 |