Learn More
STEMCELL Technologies ArciTect™ Human HPRT Positive Control Kit
Product Code. 17179301
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17179301 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17179301 Supplier STEMCELL Technologies Supplier No. 76013
 Restricted Item
Estimated Shipment: 17-11-2026
Positive control for CRISPR-Cas9 genome editing
  • ArciTect™ Human HPRT Positive Control Kit is designed to enable optimization of  transfection protocols and act as a positive control for experiments using the ArciTect™ CRISPR-Cas9 genome editing system
  • The kit comprises ArciTect™ Human HPRT crRNA (2 nmol), ArciTect™ Human HPRT Primer Mix (2 nmol), ArciTect™ tracrRNA (5 nmol), and ArciTect™ Annealing Buffer (5X)
  • ArciTect™ Human HPRT crRNA first requires annealing to ArciTect™ tracrRNA
  • This must then be combined with an ArciTect™ Cas9 Nuclease (e.g. Catalog #76002) to form a ribonucleoprotein complex
Specifications
Cell Type Pluripotent Stem Cells
Quantity 1 Kit