Learn More
Ansell Edmont™ Aramid Fiber Yellow Fabric Sleeves with Velcro Closure
Product Code. 12670962 Shop All Ansell Products
Change view
Click to view available options
Length: 559mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12670962 Length: 559mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12670962 Supplier Ansell Edmont™ Supplier No. 70123220
Only null left
Add to basket
Add to basket
Automatic knit liner of 100% Aramid fiber. Provides optimum balance of dexterity and cut resistance. Comfort and sure grip.
  • Constructed from high-tech 100% Kevlar™ Aramid fibers para-aramid short fibers.
  • Exceptional cut resistance with dexterity.
  • The 100% Dupont Kevlar™ Aramid fibers construction offers a very high cut resistance to weight ratio.
TRUSTED_SUSTAINABILITY
Specifications
Size Standard
Product Type Cut-resistant Sleeve
Material DuPont™ Aramid Fibers
Color Yellow
Sterility Non-sterile
Style Ambidextrous
Disposable Reusable
Closure Type Hook and Loop Strap
Seam Type Seamless
Length (Metric) 559 mm