missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Safety Products™ Aracut™ Grip Gloves
Click to view available options
Size:
6
7
11
Unit Size:
Each
Description
- Knitted Kevlar™ glove.
- Hand conformity and good dexterity.
- Blue PVC dots on palm for grip and abrasion resistance.
- Knitted construction for hand conformity and dexterity.
- Excellent cutting resistance and protection against thermal hazards in a dry atmosphere.
Specifications
Specifications
| Certifications/Compliance | CE CATEGORY II Intermediary risks, EN 420-94 EEC Basic standards, EN 388-94 Mechanical hazards, EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number 0075/797/162/05/08/0134, EC Attestation Number 0075/797/162/05/08/0134 |
| Coated Area | Palm, Fingertips |
| Description | Gauge 10 |
| Coating Material | PVC Dots |
| Color | Yellow |
| Cut Resistance Rating | 3 |
| Material | Aramid, Dupont™ Kevlar™ Aramid Fibers |
| Product Type | Gloves |
| Size | 6 |
| For Use With (Application) | Automotive, Building, Construction, Glass, Metal and Paper Industries |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction