missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AR-C155858

Description
AR-C155858 is an inhibitor of the monocarboxylate transporters (MCTs) MCT1 and MCT2 (Ki values are 2.3 and <10 nM respectively). Exhibits no activity at MCT4. Blocks proliferation of Raji lymphoma cells in vitro. Inhibits glycolysis and glutathione synthesis in cancer cells.
Specifications
Specifications
| Quantity | 1 mg |
| Product Type | AR-C155858 |
| Purity | >98% |
| Molecular Weight (g/mol) | 461.53 |
| Inhibitors | AR-C155858 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?