missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AR-C 66096 tetrasodium salt

Product Code. 18004332
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18004332

Brand: Tocris Bioscience™ 3321/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective P2Y12 antagonist

AR-C 66096 tetrasodium salt is a potent and selective P2Y12 receptor antagonist. Blocks ADP-induced inhibition of adenylyl cyclase in vitro (pKB =7.6) and inhibits ADP-induced aggregation of washed human platelets (pIC50 = 8.16).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Propylthio)adenosine-5'-O-(β,γ-difluoromethylene)triphosphate tetrasodium salt
CAS 145782-74-7
Quantity 1 mg
Target P2Y Receptor Antagonists
Molecular Formula C14H18F2N5Na4O12P3S
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.