missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AR-A 014418
GREENER_CHOICE

Product Code. 18064792
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18064792 50 mg 50mg
18054352 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18064792 Supplier Tocris Bioscience™ Supplier No. 3966/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective GSK-3 inhibitor

Selective glycogen synthase kinase 3 (GSK-3) inhibitor (IC50 = 104 nM). Exhibits specificity for GSK-3 over cdk2 and cdk5 (IC50 values are > 100 μM) and over 26 other kinases. Inhibits β-amyloid-mediated neurodegeneration in hippocampal slices.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 487021-52-3
Molecular Formula C12H12N4O4S

Specifications

Quantity 50 mg
Formula Weight 308.31
Purity >99%
Molecular Weight (g/mol) 308.312
Chemical Name or Material AR-A 014418
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.