missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AR-A 014418

Description
Selective glycogen synthase kinase 3 (GSK-3) inhibitor (IC50 = 104 nM). Exhibits specificity for GSK-3 over cdk2 and cdk5 (IC50 values are > 100 μM) and over 26 other kinases. Inhibits β-amyloid-mediated neurodegeneration in hippocampal slices.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 308.31 |
| Purity | >99% |
| Molecular Weight (g/mol) | 308.312 |
| Chemical Name or Material | AR-A 014418 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?