Learn More
3M™ Nomad™ Aqua Textile Drop Down Mat, Series 85
Click to view available options
Dimensions (L x W):
150 x 91.4 cm
200 x 130 cm
300 x 130 cm
300 x 200 cm
600 x 130 cm
Color:
Black
Gray
Unit Size:
Each
Description
- High performing textile entrance mat, with advanced water removal and retention properties
- Stops 80% of dirt and moisture at the door
- Entrance mat for high traffic applications (1500 to 5000 crossings per day)
- Phthalate-free, slip-resistant PVC backing
- Excellent appearance retention
- Long lasting and easy to clean
- Available as drop down mats or in rolls
Specifications
Specifications
| Dimensions (L x W) | 200 x 130 cm |
| For Use With (Application) | Heavy traffic areas |
| Model | Nomad™ Aqua, Series 85 |
| Resistance | Slip-resistant |
| Thickness (Metric) | 7.5 mm |
| Color | Black |
| Length (Metric) | 200 cm |
| Material | PVC (Backing) |
| Width (Metric) | 130 cm |