missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Aprotinin, Analytical Grade, MP Biomedicals™

Product Code. p-7083146 Shop All MP Biomedical Products
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 11485981

Brand: MP Biomedicals™ 0219038250

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Forms stable complexes with enzymes. Aprotinin, Analytical Grade, MP Biomedicals is a competitive serine proteinase inhibitor that works by blocking the active sites of enzymes. This binding is reversible with most aprotinin-protease complexes, dissociati

Aprotinin is found in bovine lymph nodes, lung, parotid gland, spleen, liver, pancreas, seminal vesicles, thyroid gland, kidney, mucous membranes of the trachea and esophagus, ovaries, heart, posterior pituitary and cartilage. It is reactive against trypsin, chymotrypsin, plasmatic and glandular kininogenases, plasmin, kallkrein, urokinase, clotting factor XIIa, protein C, proteinases of the complement system, and leukocyte and tissue proteinases. It does not inhibit thrombin. Uses include:

  • Small amounts can be added to tubes of drawn blood to enable laboratory measurement of rapidly degraded proteins, such as glucagon.
  • Can be labelled with fluorescein isothiocyanate for use as a fluorescent histochemical reagent for staining glycoconjugates (mucosubstances) that are rich in uronic or sialic acids. The conjugate retains its antiproteolytic and carbohydrate-binding properties.
  • Used in cell biology as an enzyme inhibitor to prevent protein degradation during lysis or homogenization of cells and tissues.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
CAS 9087-70-1
Synonym Pancreatic Basic Trypsin Inhibitor polypeptide; Antagosan; Antikrein; Trasylol; Zymofren; Antagosan; Anikrein; Antilysin(e); BPTI; Bayer A 128; Kallikrein-trypsin inactivator; Fosten; Iniprol; Kunitz protease inhibitor; Onquinin; Repulson; RP-9921; Ryker 52G; Triazinin; Zymofren
Content And Storage Store at 4°C
Donor Source Bovine Lung
Form Powder
For Use With (Application) Enzyme Inhibitor
Quantity 50 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.