Learn More
Miele Professional™ Appliance Lid DE-CS7-85/1
Product Code. 30533214 Shop All Miele Products
Change view
Click to view available options
Length (Metric):
90.00 cm
Unit Size:
1 sheet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
30533214 90.00 cm 1 sheet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30533214 Supplier Miele Professional™ Supplier No. 12832230
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
Cover for top of machine including side unit with 90 cm wide
  • Usable for Aqua Purificator PG 8595 and DOS cabinet PG 8596
  • Combined with 60 cm washer-disinfector (e.g. PG 8681 or PG 8682)
  • Usable for Lab Washer PLW 8683 CD
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel
Depth (English) 35 7/16 in.
Depth (Metric) 90.00 cm
Dimensions (L x W x H) 90.00 x 70.00 x 1.50 cm
Height (English) 0 5/8 in.
Height (Metric) 1.50 cm
Length (English) 35 7/16 in.
Length (Metric) 90.00 cm
Width (English) 27 9/16 in.
Width (Metric) 70.00 cm
Quantity 1 Ea.