missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ APLP2 (Human) Recombinant Protein
Human APLP2 partial ORF ( AAH00373, 41 a.a. - 150 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000334-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: AGTGFAVAEPQIAMFCGKLNMHVNIQTGKWEPDPTGTKSCFETKEEVLQYCQEMYPELQITNVMEANQRVSIDNWCRRDKKQCKSRFVTPFKCLVGEFVSDVLLVPEKCQ
Specifications
AAH00373 | |
amyloid beta (A4) precursor-like protein 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
APLP2 | |
GST |
334 | |
Wheat germ expression system | |
25 μg | |
APPH, APPL2, CDEBP | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |