missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ APEX1 (Human) Recombinant Protein
Human APEX1 full-length ORF ( AAH02338, 1 a.a. - 318 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000328-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MPKRGKKGAVAEDGDELRTEPEAKKSKTAAKKNDKEAAGEGPALYEDPPDQKTSPSGKPATLKICSWNVDGLRAWIKKKGLDWVKEEAPDILCLQETKCSENKLPAELQELPGLSHQYWSAPSDKEGYSGVGLLSRQCPLKVSYGIGEEEHDQEGRVIVAEFDSFVLVTAYVPNAGRGLVRLEYRQRWDEAFRKFLKGLASRKPLVLCGDLNVAHEEIDLRNPKGNKKNAGFTPQERQGFGELLQAVPLADSFRHLYPNTPYAYTFWTYMMNARSKNVGWRLDYFLLSHSLLPALCDSKIRSKALGSDHCPITLYLAL
Specifications
AAH02338 | |
APEX nuclease (multifunctional DNA repair enzyme) 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
APEX1 | |
GST |
328 | |
Wheat germ expression system | |
10 μg | |
APE, APE-1, APE1, APEN, APEX, APX, HAP1, REF-1, REF1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |