Learn More
mb2 Polyurethane Stool on Castors - Round
Product Code. 15744308
Click to view available options
:
Medium Height, Casters
Product Code. 15744308
Brand: mb2 AP6B2D1NE2
Only null left
Add to basket
Add to basket
Polyurethane stool on castors - Round - Mid lift - Fix mechanism
  • Stool in polyurethane, round
  • Seat in polyurethane
  • Pneumatic cylinder mid
  • 5-spoke base injected in nylon and fiberglass reinforced by a galvanized steel ring.
  • Hard floor castors 50 mm
Specifications
Product Type Stool
Color Black
Adjustable (Height) Yes
Adjustable Height
Autoclavable Not Autoclavable
Frame Material Steel
Includes Five-star nylon and fiberglass base. Casters.
Quantity 1 Ea.
Seat Material Polyurethane
Seating Height 470 to 650 mm
Warranty 5 Years
Weight (Metric) 6.06 kg
With Armrest No
With Backrest No
With Casters Yes
Footrest No
With Lumbar Support No
With Tilt No
Material Polyurethane
Seating Type Stool