missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Product Line | Novus |
| Target | AP3M2 |
| Synonym | Adapter-related protein complex 3 mu-2 subunit, adaptor-related protein complex 3, mu 2 subunit, AP-3 complex subunit mu-2, AP47B, CLA20, Clathrin assembly protein assembly protein complex 1 medium chain homolog 2, Clathrin coat assembly protein AP47 homolog 2, Clathrin coat-associated protein AP47 homolog 2, clathrin-associated protein AP47 homolog 2, Golgi adaptor AP-1 47 kDa protein homolog 2, HA1 47 kDa subunit homolog 2, HA1 47kDA subunit homolog 2, mu3B-adaptin, P47B |
| Gene Symbol | AP3M2 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?