missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ANXA2 (Human) Recombinant Protein

Human ANXA2 full-length ORF ( AAH09564.1, 1 a.a. - 339 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00000302-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16121611

  • 545.00€ / 25µg

Estimated Shipment: 16-09-2022
to see stock.
Add to basket

Description

Description

  • Sequence: MFTVHEILCKLSLEGDHSTPPSAYGSVKAYTNFDAERDALNIETAIKTKGVDEVTIVNILTNRSNAQRQDIAFAYQRRTKKELASALKSALSGHLETLILGLLKTPAQYDASELKASMKGLGTDEDSLIEIICSRTNQELQEINRVYKEMYKTDLEKDIISDTSGDFRKLMVALAKGRRAEDGSVIDYELIDQDARDLYDAGVKRKGTDVPKWISIMTERSVPHLQKVFDRYKSYSPYDMLESIRKEVKGDLENAFLNLVQCIQNKPLYFADRLYDSMKGKGTRDKVLIRIMVSRSEVDMLKIRSEFKRKYGKSLYYYIQQDTKGDYQKALLYLCGGDD
Specifications

Specifications

AAH09564.1
annexin A2
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ANXA2
GST
302
Wheat germ expression system
25 μg
ANX2, ANX2L4, CAL1H, LIP2, LPC2, LPC2D, P36, PAP-IV
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents
Certificates
Special Offers

Special Offers