missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ANXA10 Recombinant Protein

Brand:  Abnova™ H00011199-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16116362

  • 508.00€ / 25µg
Estimated Shipment:
to see stock.
Add to basket

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH07320
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
61.38
8
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ANXA10
Human
Recombinant
Solution
Antibody Production, Protein Array, ELISA, Western Blot
11199
ANXA10 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
ANX14
ANXA10
Wheat Germ (in vitro)
GST
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers