missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 mg |
| Formula Weight | 604.7 |
| Physical Form | Powder |
| Chemical Name or Material | Antipain |
Safety and Handling
Product Identifier
- Antipain Hydrochloride
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Antipain Hydrochloride
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?