missing translation for 'onlineSavingsMsg'
Learn More

Antipain, MP Biomedicals™

Product Code. 11473991 Shop All MP Biomedical Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473991 5 mg 5mg
11483991 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11473991 Supplier MP Biomedicals Supplier No. 0215284305

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 37691-11-5
Molecular Formula C27H44N10O6
Molecular Weight (g/mol) 604.71
MDL Number MFCD00135959
InChI Key SDNYTAYICBFYFH-TUFLPTIASA-N
Synonym antipain, antipain hydrochloride dihydrate, antipain, >50000 u/mg, s-1-carboxy-2-phenylethyl carbamoyl-l-arginyl-l-valyl-argininal, 2s-2-1s-4-carbamimidamido-1-1s-1-5-carbamimidamido-1-oxopentan-2-yl carbamoyl-2-methylpropyl carbamoyl butyl carbamoyl amino-3-phenylpropanoic acid
PubChem CID 133126507
IUPAC Name 2-[[(2R)-5-(diaminomethylideneamino)-1-[[1-[[5-(diaminomethylideneamino)-1-oxopentan-2-yl]amino]-3-methyl-1-oxobutan-2-yl]amino]-1-oxopentan-2-yl]carbamoylamino]-3-phenylpropanoic acid
SMILES CC(C)[C@H](NC(=O)[C@H](CCCN=C(N)N)NC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O)C(=O)N[C@@H](CCCN=C(N)N)C=O

Specifications

Color Yellow
Quantity 5 mg
Formula Weight 604.7
Physical Form Powder
Chemical Name or Material Antipain
compliance-icons
Product Identifier
  • Antipain Hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Antipain Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.