Learn More
anti-VAMP3/Cellubrevin, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand:  Novus Biologicals NB300-510
Encompass
Product Code. 12831976
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
VAMP3/Cellubrevin Polyclonal specifically detects VAMP3/Cellubrevin in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.