missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta RII, Goat anti-Human, FITC, Polyclonal, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems FAB2411F-025
This item is not returnable.
View return policy
Description
TGF-beta RII Polyclonal specifically detects TGF-beta RII in Human samples. It is validated for Flow Cytometry.Specifications
| TGF-beta RII | |
| Polyclonal | |
| Flow Cytometry 10 uL/10^6 cells | |
| P37173.2 | |
| TGFBR2 | |
| Mouse myeloma cell line NS0-derived recombinant human TGF-beta RII Ile24-Asp159 Accession # P37173.2 | |
| 25 Tests | |
| Primary | |
| Detects human TGF-beta RII in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse (rm) TGF-beta RII and less than 1% cross-reactivity with recombinant human TGF-beta RIII and rmTGF-beta RI is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| Flow Cytometry | |
| Fluorescein | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| AAT3, EC 2.7.11, EC 2.7.11.30, FAA3, HNPCC6, LDS1B, LDS2B, MFS2, RIIC, TAAD2, tbetaR-II, TGF-beta receptor type II, TGF-beta receptor type IIB, TGF-beta receptor type-2, TGF-beta type II receptor, TGFbetaRII, TGFbeta-RII, TGFBR2, TGF-bRII, TGFR-2, transforming growth factor beta receptor type IIC, transforming growth factor, beta receptor II (70/80kDa), transforming growth factor, beta receptor II (70/80kDa) isoform 1, transforming growth factor, beta receptor II (70/80kDa) isoform 2, transforming growth factor, beta receptor II (70-80kD), Transforming growth factor-beta receptor type II | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7048 | |
| Human | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?