Learn More
BD Pharmingen™ PE-Cy™7 Streptavidin
Click to view available options
Quantity:
0.1 mg
Unit Size:
Each
Description
- Flow Cytometry
Specifications
Specifications
| Antigen | Streptavidin |
| Concentration | 0.2 mg/mL |
| Conjugate | PE-Cy7 |
| Quantity | 0.1 mg |
| Regulatory Status | For research use only (RUO) |
| Storage Buffer | Aqueous buffered solution containing less than or equal to 0.09% sodium azide |
| Content And Storage | Store undiluted at 4°C. Protect from prolonged exposure to light. Do not freeze. |
| For Use With (Application) | Flow Cytometry |