Learn More
Staphylococcus Enterotoxin A Monoclonal Antibody (J98J), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA17481
Additional Details : Weight : 0.01000kg
Description
The MA1-7481 antibody reacts with Staphylococcus Enterotoxin toxin A probe in bacterial and infected tissue samples. See related products for paired antibody. MA1-7481 has been successfully used in ELISA applications. The MA1-7481 antibody was raised against Staphylococcus Toxin A.
Staphylococcus Enterotoxin A Monoclonal antibody specifically detects Staphylococcus Enterotoxin A in Bacteria samples. It is validated for ELISA.Specifications
Staphylococcus Enterotoxin A | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Staph. Toxin A. | |
100 μg | |
Primary | |
4° C | |
Liquid |
ELISA | |
J98J | |
Unconjugated | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG1 |