missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCP1 Rabbit anti-Mouse, Rat, Chicken, Mammalia, Parasite, Monkey, Human (Negative), Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 45 publications
Brand: Novus Biologicals NB300-229
This item is not returnable.
View return policy
Description
SCP1 Polyclonal specifically detects SCP1 in Mouse, Rat, Chicken, Mammal, Parasite, Monkey, Human (Negative) samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| SCP1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| SYCP1 | |
| A synthetic peptide made to the C-terminus of the mouse SCP1 protein sequence. [UniProt# Q62209] | |
| 0.1 mL | |
| Primary | |
| Mouse, Rat, Chicken, Mammalia, Parasite, Monkey, Human (Negative) | |
| IgG |
| Western Blot, ChIP assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 1.0 mg/mL | |
| Western Blot, Simple Western 1:100, Chromatin Immunoprecipitation 1:10-1:500, Immunohistochemistry 1:750, Immunocytochemistry/Immunofluorescence 1:100-1:750, Immunoprecipitation, Immunohistochemistry-Paraffin 1:750, Immunohistochemistry-Frozen 1:750 | |
| Cancer/testis antigen 8, CT8MGC104417, HOM-TES-14, SCP1SCP-1, synaptonemal complex protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 6847 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only