Learn More
anti-S100 calcium binding protein A14, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NBP1-90000
Encompass
Product Code. 15785166
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
S100 calcium binding protein A14 Polyclonal specifically detects S100 calcium binding protein A14 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.