Learn More
anti-RUNX2/CBFA1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP2-24755SS
Description
RUNX2/CBFA1 Polyclonal specifically detects RUNX2/CBFA1 in Human, Mouse, Chicken, Equine, Primate samples. It is validated for Western Blot.Specifications
| RUNX2/CBFA1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 0.05% BSA with 0.05% Sodium Azide | |
| Acute myeloid leukemia 3 protein, AML3CCD, CBFA1MGC120022, Core-binding factor subunit alpha-1, MGC120023, PEA2-alpha A, PEBP2A, PEBP2A1CCD1, PEBP2aA1, PEBP2-alpha A, runt domain, alpha subunit 1, runt-related transcription factor 2, SL3/AKV core-binding factor alpha A subunit, SL3-3 enhancer factor 1 alpha A subunit | |
| Rabbit | |
| 56.6 kDa | |
| 0.025 mg | |
| Primary | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in human, rhesus monkey, macaque, chimpanzee, orangutan, gorilla, cow, bison, horse, opossum and boar, and 90% homologous in dog and rat. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/mL | |
| Q13950 | |
| RUNX2 | |
| This RUNX2/CBFA1 Antibody was developed against a synthetic peptide corresponding to somewhere between amino acids 250-300 of human RUNX2 was used as immunogen for RUNX2/CBFA1 Antibody. RUNX2 and RUNX1 share an approximate 66% homology in peptide sequence used as immunogen. | |
| Protein G purified | |
| RUO | |
| 860 | |
| Human, Mouse, Chicken, Equine, Primate | |
| Purified |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only