missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Rat IgG2a, kappa Isotype Control, Clone: R35-95, BD
Brand: BD Biosciences 563335
Additional Details : Weight : 0.02000kg
Description
Host Species: Rat
Clone: R35-95
Isotype: IgG2a, kappa
Species Reactivity: Hu, Ms
Immunogen: Mouse Pooled Immunoglobulin
Isotype Control
Specifications
Rat IgG2a, kappa | |
R35-95 | |
Brilliant Violet 786 | |
Affinity Purified | |
RUO | |
Human, Mouse |
Monoclonal | |
0.2mg/mL | |
Rat | |
50 μg | |
Secondary | |
IgG2a κ |