missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMA1 Monoclonal Antibody (40B7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA191567
Additional Details : Weight : 0.01000kg
Description
The predicted molecular weight by western blot is 100 kDa.
PMA1 Monoclonal antibody specifically detects PMA1 in Yeast samples. It is validated for Immunocytochemistry, Western Blot.Specifications
PMA1 | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with 10mM sodium azide | |
P05030 | |
PMA1 | |
Yeast membrane preps. | |
RUO | |
852876 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
40B7 | |
Unconjugated | |
PMA1 | |
H(+)-exporting P2-type ATPase PMA1; KTI10; Plasma membrane ATPase 1; PMA1; Proton pump 1; YGL008C | |
Mouse | |
250 μL | |
Primary | |
Yeast | |
Antibody | |
IgG |