missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLK4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529373
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: PLK4-transfected 293T. Predicted reactivity: Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PLK4 Polyclonal antibody specifically detects PLK4 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
PLK4 | |
Polyclonal | |
Unconjugated | |
PLK4 | |
1700028H20; AI385771; MCCRP2; PLK4; PLK-4; polo like kinase 4; polo-like kinase 4; SAK; serine/threonine kinase 18; serine/threonine-protein kinase 18; Serine/threonine-protein kinase PLK4; serine/threonine-protein kinase Sak; Snk akin kinase; Stk18 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10733 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.19 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
O00444 | |
PLK4 | |
Recombinant fragment corresponding to a region within amino acids 614 and 871 of Human PLK4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |