Learn More
Pancreatic Polypeptide/PP Goat anti-Human, Mouse, Monkey, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody has been used in 16 publications
Brand: Novus Biologicals NB100-1793
Description
Pancreatic Polypeptide/PP Polyclonal specifically detects Pancreatic Polypeptide/PP in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Peptide ELISA.Specifications
| Pancreatic Polypeptide/PP | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| Obinepitide, pancreatic polypeptide (ppy), pancreatic polypeptide PNPPP, pancreatic polypeptide Y, pancreatic prohormone | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5539 | |
| Human, Mouse, Monkey | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, ELISA | |
| 0.5 mg/mL | |
| Flow Cytometry, Immunohistochemistry 5 ug/mL, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 3 ug/ml, Immunohistochemistry-Frozen 1:500, Immunoblotting, Peptide ELISA Detection limit 1:32000 | |
| P01298 | |
| PPY | |
| Peptide with sequence C-TRPRYGKRHKEDT corresponding to internal region according to NP_002713.1. | |
| 0.1 mg | |
| Primary | |
| Expected cross-reactivity based on sequence identity: Rat, Dog. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.
For Research Use Only