Learn More
anti-XPC, Clone: 3.26, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 17 publications
Brand:  Novus Biologicals NB100-477
Encompass
Product Code. 15737315
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
XPC Monoclonal specifically detects XPC in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.