Learn More
Novus Biologicals™ Anti-Mouse Ig (H+L) Comp-Bead 3 Population (7.5 um) Kit
Product Code. 18355146 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
3 x 5 mL
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18355146 3 x 5 mL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18355146 Supplier Novus Biologicals™ Supplier No. NBP300499
  • ONLINE EXCLUSIVE PRICE
    481.00€
    454.65€ / 1 sets
    Save 26.35€
    5% Off
 Restricted Item
Estimated Shipment: 06-10-2026
The Anti-Mouse Ig (H+L) Compensation Beads are designed to capture fluorophore-conjugated antibodies, serving as accurate and consistent compensation controls in multicolor flow cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at 4°C. Do not freeze.
Quantity 3 x 5 mL
Formulation PBS, 0.2% BSA
For Use With (Application) Flow Cytometry