Learn More
Novus Biologicals™ Anti-Mouse Ig (H+L) Comp-Bead 3 Population (5.5 um) Kit
Shop All Bio Techne ProductsSpecifications
Specifications
| Content And Storage | Store at 4°C in the dark. Do not freeze. |
| Quantity | 3 x 5 mL |
| Formulation | PBS, 0.2% BSA |
| For Use With (Application) | Flow Cytometry |