Learn More
anti-DUX4, Clone: P4H2, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NBP1-49552
Description
DUX4 Monoclonal specifically detects DUX4 in Human samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| DUX4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 0.5-2.0ug/ml, Simple Western 1:10, ELISA, Immunohistochemistry 1:100-1:200, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:100-1:200, Immunohistochemistry-Frozen 1:100-1:200 | |
| Q9UBX2 | |
| DUX4 | |
| Synthetic peptide corresponding to the C-terminus of human DUX4. [Swiss-Prot: Q9UBX2] | |
| Protein G purified | |
| RUO | |
| 100288687 | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| P4H2 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| double homeobox 4, double homeobox protein 10, double homeobox protein 4, double homeobox protein 4/10, double homeobox protein DUX10, DUX10 | |
| Mouse | |
| 52 kDa | |
| 0.1 mL | |
| Primary | |
| This antibody is specific for DUX4 and does not recognize DUX4c. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only