Learn More
beta Amyloid Mouse anti-Human, Mouse, Rat, Bacteria, Monkey, Clone: MOAB-2, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 34 publications
Brand: Novus Biologicals NBP2-13075
Description
beta Amyloid Monoclonal specifically detects beta Amyloid in Human, Mouse, Rat, Bacteria, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Dot Blot, Immunohistochemistry Free-Floating.Specifications
| beta Amyloid | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:1000-1:5000, ELISA 1:100-1:1000, Immunohistochemistry 1:40-1:400, Immunocytochemistry/Immunofluorescence 1:200-1:500, Immunoprecipitation 1:200-1:1000, Immunohistochemistry-Paraffin 1:40-1:400, Immunohistochemistry-Frozen, Immunoblotting, Dot Blot, Immunohistochemistry Free-Floating | |
| P05067 | |
| APP | |
| This beta Amyloid antibody was developed against recombinant human beta Amyloid 42. | |
| Protein G purified | |
| RUO | |
| 351 | |
| Human, Mouse, Rat, Bacteria, Monkey | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| MOAB-2 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| Abeta, Amyloid beta, beta-amyloid peptide | |
| Mouse | |
| 5 kDa | |
| 0.1 mL | |
| Primary | |
| Beta Amyloid antibody (MOAB-2) recognizes unaggregated, oligomeric and fibrillar forms of beta Amyloid 42 and unaggregated beta Amyloid 40. Does not detect APP. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only