Learn More
anti-Ataxin 1, Clone: 2F5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP1-51689
Encompass
Product Code. 15708525
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Ataxin 1 Monoclonal specifically detects Ataxin 1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.