Learn More
anti-AMPK alpha 1, Clone: 2B7, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 9 publications
Brand: Novus Biologicals NBP2-22127
Description
AMPK alpha 1 Monoclonal specifically detects AMPK alpha 1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| AMPK alpha 1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1 - 2 μg/mL, Simple Western 1:50, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 2 - 4 μg/mL, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Q13131 | |
| PRKAA1 | |
| Purified recombinant fragment of human AMPK alpha 1 expressed in E. coli. [UniProt# Q13131] | |
| Protein A or G purified | |
| RUO | |
| 5562 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 2B7 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| AMP -activate kinase alpha 1 subunit, AMP-activated protein kinase, catalytic, alpha-1,5'-AMP-activated protein kinase catalytic subunit alpha-1, AMPK, AMPK alpha 1,5'-AMP-activated protein kinase, catalytic alpha-1 chain, AMPK subunit alpha-1, AMPK1, AMPKa1, EC 2.7.11, EC 2.7.11.1, MGC33776, MGC57364, protein kinase, AMP-activated, alpha 1 catalytic subunit | |
| Mouse | |
| 64 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Primate | |
| Precipitation |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only