Learn More
Ly-6G Rat anti-Mouse, APC-Cy7, Clone: 1A8, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 560600
Additional Details : Weight : 0.01000kg
Description
The 1A8 antibody reacts with Ly-6G, a 21 to 25kDa GPI-anchored protein. In the bone marrow, Ly6G is expressed on the majority of the largest cells, which are predominantly granulocytes, and not on lymphoid or erythroid cells. In the periphery, it is expre
Host Species: Rat
Clone: 1A8
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Ly-6G-transfected EL4J Cell Line
Flow Cytometry
Specifications
Ly-6G | |
Monoclonal | |
0.2mg/mL | |
Ly6g; Lymphocyte antigen 6G; Lymphocyte antigen 6 complex, locus G; Gr1 | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
1A8 | |
APC-Cyanine7 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Ly-6G-transfected EL4J Cell Line | |
50 μg | |
Immunology | |
Mouse | |
IgG2a κ |