Learn More
anti-LOX propeptide, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NBP1-30327
Description
LOX propeptide Polyclonal specifically detects LOX propeptide in Human, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| LOX propeptide | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| lysyl oxidaseEC 1.4.3.13, MGC105112, protein-lysine 6-oxidase | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 4015 | |
| Human, Rat | |
| IgG |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 1 mg/mL | |
| Western Blot 0.5 ug/ml, Simple Western 1:200, Flow Cytometry 1:200, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:10-1:500 | |
| P28300 | |
| LOX | |
| Synthetic peptide made to an internal portion of human LOX propeptide (within residues 118-168). [Swiss-Prot# P28300] | |
| 0.1 mL | |
| Primary | |
| This antibody recognizes the glycosylated propeptide form (∼35 kDa) and the proenzyme form (∼50 kDa). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only