Learn More
anti-IgG2b κ, Rat, APC-Cy™7, Clone: A95-1, Isotype Control, BD
Brand: BD Biosciences 552773
Additional Details : Weight : 0.01000kg
Description
The A95-1 antibody has unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten that is not expressed on human, mouse, rat, or non-human primate cells. The A95-1 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only.
Host Species: Rat
Clone: A95-1
Isotype: IgG2b κ
Immunogen: TNP-Keyhole Limpet Hemocyanin
Flow Cytometry, Isotype Control
Specifications
Flow Cytometry | |
0.2mg/mL | |
Rat | |
Affinity Purified | |
Undiluted at 4°C. Protect from exposure to light. Do not freeze. |
A95-1 | |
APC-Cyanine7 | |
TNP-keyhole limpet hemocyanin | |
0.1 mg | |
IgG2b κ |