missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IgG2a λ, Rat, PE, Clone: B39-4, BD
Brand: BD Biosciences 557076
Additional Details : Weight : 0.01000kg
Description
The B39-4 antibody is specific for keyhole limpet hemocyanin (KLH).
Host Species: Rat
Clone: B39-4
Isotype: IgG2a, <special id='310'/>Immunogen: Trinitrophenol-KLH
Flow Cytometry, Isotype Control
Specifications
Flow Cytometry | |
0.2mg/mL | |
Rat | |
Affinity Purified | |
Undiluted at 4°C |
B39-4 | |
PE | |
Trinitrophenol-KLH | |
0.1 mg | |
IgG2a |