missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IgG1 κ, Isotype Control, Clone: A19-3, BD
Brand: BD Biosciences 563009
Additional Details : Weight : 0.01000kg
Description
Host Species: Hamster
Clone:A19-3
IsotypeIgG1, κ
Immunogen:TNP-keyhole limpet hemocyanin (KLH)
Flow Cytometry, Isotype Control
Specifications
Flow Cytometry | |
A19-3 | |
Brilliant Violet 605 | |
TNP-keyhole limpet hemocyanin | |
50 μg | |
Isotype Control (Primary) | |
IgG1 κ |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |