Learn More
anti-IgG1 κ, Hamster, Alexa Fluor™ 488, Clone: A19-3, BD
Brand: BD Biosciences 557674
Additional Details : Weight : 0.01000kg
Description
The A19-3 clone reacts with the hapten, trinitrophenol (TNP). TNP is not expressed on human, mouse, or rat cells. The immunoglobulin from clone A19-3 was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only.
Host Species: Hamster
Clone: A19-3
Isotype: IgG1 κImmunogen: TNP-keyhole limpet hemocyanin
Flow Cytometry, Isotype Control
Specifications
Flow Cytometry | |
0.2mg/mL | |
Hamster | |
Affinity Purified | |
Undiluted at 4°C |
A19-3 | |
Alexa Fluor 488 | |
TNP-keyhole limpet hemocyanin | |
0.1 mg | |
IgG1 κ |